missing translation for 'onlineSavingsMsg'
Learn More

MCP-2 Polyclonal Antibody, Biotin, PeproTech®, Invitrogen™

Rabbit Polyclonal Antibody

Brand:  Invitrogen 500-P35BT-50UG

 View more versions of this product

Product Code. 17844883

  • £212.00 / 50µg

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

AA Sequence of recombinant protein: QPDSVSIPIT CCFNVINRKI PIQRLESYTR ITNIQCPKEA VIFKTKRGKE VCADPKERWV RDSMKHLDQI FQNLKP. Preparation: Produced from sera of rabbits immunized with highly pure Recombinant Human MCP-2 (CCL8). Anti-Human MCP-2 (CCL8)-specific antibody was purified by affinity chromatography and then biotinylated. Sandwich ELISA: To detect hMCP-2 by sandwich ELISA (using 100 μL/well antibody solution) a concentration of 0.25-1.0 μg/mL of this antibody is required. This biotinylated polyclonal antibody, in conjunction with PeproTech Polyclonal Anti-Human MCP-2 (500-P35) as a capture antibody, allows the detection of at least 0.2-0.4 ng/well of Recombinant hMCP-2. Western Blot: To detect hMCP-2 by Western Blot analysis this antibody can be used at a concentration of 0.1-0.2 μg/mL. Used in conjunction with compatible secondary reagents the detection limit for Recombinant hMCP-2 is 1.5-3.0 ng/lane, under either reducing or non-reducing conditions. 500-P35BT-1MG will be provided as 2 x 500 μg

This antimicrobial gene is one of several chemokine genes clustered on the q-arm of chromosome 17. Chemokines form a superfamily of secreted proteins involved in immunoregulatory and inflammatory processes. The superfamily is divided into four subfamilies based on the arrangement of N-terminal cysteine residues of the mature peptide. This chemokine is a member of the CC subfamily which is characterized by two adjacent cysteine residues. This cytokine displays chemotactic activity for monocytes, lymphocytes, basophils and eosinophils. By recruiting leukocytes to sites of inflammation this cytokine may contribute to tumor-associated leukocyte infiltration and to the antiviral state against HIV infection.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

MCP-2
Polyclonal
Biotin
CCL8
1810063B20Rik; AB023418; C-C motif chemokine 8; C-C motif chemokine ligand 8; CCL8; chemokine (C-C motif) ligand 8; HC14; MCP2; MCP-2; MCP-2(6-76); monocyte chemoattractant protein 2; monocyte chemoattractant protein-2; monocyte chemotactic protein 2; SCYA10; SCYA8; small inducible cytokine A8; small inducible cytokine subfamily A (Cys-Cys), member 8 (monocyte chemotactic protein 2); small-inducible cytokine A8
Rabbit
Antigen affinity chromatography
RUO
6355
-20°C
ELISA, Western Blot
0.1-1.0 mg/mL
PBS with no preservative
P80075
CCL8
E.coli-derived Recombinant Human MCP-2 (CCL8)
50 μg
Primary
Human
Lyophilized
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers