Learn More
MCP-2 Polyclonal Antibody, PeproTech®, Invitrogen™
Rabbit Polyclonal Antibody
£165.00 - £1374.00
Specifications
Antigen | MCP-2 |
---|---|
Concentration | 0.1-1.0 mg/mL |
Applications | ELISA, Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
17833853
|
Invitrogen
500-P127-50UG |
50μg |
£165.00
50µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
17844363
|
Invitrogen
500-P127-100UG |
100 μg |
£212.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
17824783
|
Invitrogen
500-P127-1MG |
1 mg |
£1374.00
1mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
AA Sequence of recombinant protein: GPDKAPVTCC FHVLKLKIPL RVLKSYERIN NIQCPMEAVV FQTKQGMSLC VDPTQKWVSE YMEILDQKSQ ILQP. Preparation: Produced from sera of rabbits immunized with highly pure Recombinant Murine MCP-2 (CCL8). Anti-Murine MCP-2 (CCL8)-specific antibody was purified by affinity chromatography employing an immobilized Murine MCP-2 (CCL8) matrix. Sandwich ELISA: To detect Murine MCP-2 (CCL8) by sandwich ELISA (using 100 μL/well antibody solution) a concentration of 0.5-2.0 μg/mL of this antibody is required. This antigen affinity purified antibody, in conjunction with PeproTech Biotinylated Anti-Murine MCP-2 (CCL8) (500-P127Bt) as a detection antibody, allows the detection of at least 0.2-0.4 ng/well of Recombinant Murine MCP-2 (CCL8). Western Blot: To detect Murine MCP-2 (CCL8) by Western Blot analysis this antibody can be used at a concentration of 0.1-0.2 μg/mL. Used in conjunction with compatible secondary reagents the detection limit for Recombinant Murine MCP-2 (CCL8) is 1.5-3.0 ng/lane, under either reducing or non-reducing conditions.
This antimicrobial gene is one of several chemokine genes clustered on the q-arm of chromosome 17. Chemokines form a superfamily of secreted proteins involved in immunoregulatory and inflammatory processes. The superfamily is divided into four subfamilies based on the arrangement of N-terminal cysteine residues of the mature peptide. This chemokine is a member of the CC subfamily which is characterized by two adjacent cysteine residues. This cytokine displays chemotactic activity for monocytes, lymphocytes, basophils and eosinophils. By recruiting leukocytes to sites of inflammation this cytokine may contribute to tumor-associated leukocyte infiltration and to the antiviral state against HIV infection.Specifications
MCP-2 | |
ELISA, Western Blot | |
Unconjugated | |
Rabbit | |
Mouse | |
Q9Z121 | |
20307 | |
E.coli-derived Recombinant Murine MCP-2 (CCL8) | |
Antigen affinity chromatography | |
CCL8 |
0.1-1.0 mg/mL | |
Polyclonal | |
Lyophilized | |
RUO | |
PBS with no preservative | |
1810063B20Rik; AB023418; C-C motif chemokine 8; C-C motif chemokine ligand 8; CCL8; chemokine (C-C motif) ligand 8; HC14; MCP2; MCP-2; MCP-2(6-76); monocyte chemoattractant protein 2; monocyte chemoattractant protein-2; monocyte chemotactic protein 2; SCYA10; SCYA8; small inducible cytokine A8; small inducible cytokine subfamily A (Cys-Cys), member 8 (monocyte chemotactic protein 2); small-inducible cytokine A8 | |
CCL8 | |
Primary | |
-20°C |