missing translation for 'onlineSavingsMsg'
Learn More

MCP-4 Polyclonal Antibody, PeproTech®, Invitrogen™

Goat Polyclonal Antibody

£165.00 - £1374.00

Specifications

Antigen MCP-4
Concentration 0.1-1.0 mg/mL
Applications ELISA, Western Blot
Classification Polyclonal
Conjugate Unconjugated
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
17893823
View Documents Promotion Details
Invitrogen
500-P04G-50UG
50μg
£165.00
50µg
Please to purchase this item. Need a web account? Register with us today!
17814043
View Documents Promotion Details
Invitrogen
500-P04G-100UG
100 μg
£212.00
100µg
Please to purchase this item. Need a web account? Register with us today!
17843913
View Documents Promotion Details
Invitrogen
500-P04G-1MG
1 mg
£1374.00
1mg
Please to purchase this item. Need a web account? Register with us today!
Description

Description

AA Sequence of recombinant protein: QPDALNVPST CCFTFSSKKI SLQRLKSYVI TTSRCPQKAV IFRTKLGKEI CADPKEKWVQ NYMKHLGRKA HTLKT. Preparation: Produced from sera of goats immunized with highly pure Recombinant Human MCP-4 (CCL13). Anti-Human MCP-4 (CCL13)-specific antibody was purified by affinity chromatography employing an immobilized Human MCP-4 matrix. Sandwich ELISA: To detect Human MCP-4 (CCL13) by sandwich ELISA (using 100 μL/well) a concentration of 0.5-2.0 μg/mL of this antibody is required. This antigen affinity purified antibody, in conjunction with PeproTech Biotinylated Anti-Human MCP-4 (CCL13) (500-P04GBT) as a detection antibody, allows the detection of at least 2000- 4000 pg/mL of Recombinant Human MCP-4 (CCL13). Western Blot: To detect Human MCP-4 (CCL13) by Western Blot analysis this antibody can be used at a concentration of 0.1-0.2 μg/mL. When used in conjunction with compatible secondary reagents the detection limit for Recombinant Human MCP-4 (CCL13) is 1.5-3.0 ng/lane, under either reducing or non-reducing conditions.

Monocyte Chemotactic Protein 4 (MCP-4), also called CCL13, is induced by inflammatory proteins such as IL-1 and TNFalpha. MCP-4 is a ligand for three different G protein coupled receptors, CCR2, CCR3 and CCR5. MCP-4 activates signaling in monocytes, T lymphocytes, eosinophils and basophils and this signaling is associated with the allergic response.
Specifications

Specifications

MCP-4
ELISA, Western Blot
Unconjugated
Goat
Human
Q99616
6357
E.coli-derived Recombinant Human MCP-4 (CCL13)
Antigen affinity chromatography
CCL13
0.1-1.0 mg/mL
Polyclonal
Lyophilized
RUO
PBS with no preservative
C-C motif chemokine 13; C-C motif chemokine 13, long chain; C-C motif chemokine 13, medium chain; C-C motif chemokine 13, short chain; C-C motif chemokine ligand 13; CCL 13; CCL13; chemokine (C-C motif) ligand 13; CKb10; CK-beta-10; H-MCP-4; MCP4; MCP-4; Monocyte chemoattractant protein 4; monocyte chemotactic protein 4; NCC1; NCC-1; new CC chemokine 1; SCYA13; SCYL1; small inducible cytokine subfamily A (Cys-Cys), member 13; small-inducible cytokine A13
CCL13
Primary
-20°C
Videos
SDS
Documents

Documents

Product Certifications