Learn More
Invitrogen™ MCP-5 (CCL12) Mouse ELISA Kit
Sandwich ELISA Kit
Brand: Invitrogen™ EMCCL12X5
Description
The Mouse MCP-5 (CCL12) ELISA quantitates Ms MCP-5 in mouse serum, plasma, or cell culture medium. The assay will exclusively recognize both natural and recombinant Ms MCP-5. Principle of the method The Mouse MCP-5 solid-phase sandwich ELISA (enzyme-linked immunosorbent assay) is designed to measure the amount of the target bound between a matched antibody pair. A target-specific antibody has been pre-coated in the wells of the supplied microplate. Samples, standards, or controls are then added into these wells and bind to the immobilized (capture) antibody. The sandwich is formed by the addition of the second (detector) antibody, a substrate solution is added that reacts with the enzyme-antibody-target complex to produce measurable signal. The intensity of this signal is directly proportional to the concentration of target present in the original specimen. Rigorous validation Each manufactured lot of this ELISA kit is quality tested for criteria such as sensitivity, specificity, precision, and lot-to-lot consistency. See manual for more information on validation.
Chemotactic factor that attracts eosinophils, monocytes, and lymphocytes but not neutrophils. Potent monocyte active chemokine that signals through CCR2. Involved in allergic inflammation and the host response to pathogens and may play a pivotal role during early stages of allergic lung inflammation.Specifications
Q62401 | |
0.5 pg/mL | |
Solid-phase sandwich ELISA | |
ELISA Kit | |
Plasma, Serum, Supernatant | |
5 x 96 assays | |
ELISA, Protein Assays, Protein Biology, Cancer Biology, Immunology, Neuroscience, Neurobiology, Protein Detection, Protein Analysis | |
20293 | |
<12% | |
Standard, Pre-coated 96 well plates, Adhesive Plate Covers, Wash Buffer, Chromogen, Biotinylated Detection Antibody, SAV-HRP, Stop Solution, Assay Diluent Concentrate | |
5 x 96 Tests | |
Angiogenesis | |
2°C to 8°C | |
Mouse | |
4 hr. 45 min. |
0.41 pg/mL to 100 pg/mL | |
0.5 pg/mL to 100 pg/mL | |
Biotin | |
Chemokine & Receptors | |
This ELISA kit shows no cross-reactivity with any of the cytokines tested: Mouse 6 Ckine, CTACK, Eotaxin, GCSF, GM-CSF, IL-2, IL-3, IL-4, IL-5, IL-6, IL-9, IL-10, IL-12p40, IL-12p70, IL-13, IL-17, IFN-gamma, KC, Leptin, MCP-1, MIP-1 alpha, MIP-2, MIP-3 beta, RANTES, SCF, sTNFri, TARC, TIMP-1, TNF-alpha, TPO, VEGF. | |
Mouse | |
Colorimetric Microplate Reader | |
MCP-5,Scya12 | |
<10% | |
HRP | |
RUO | |
Plasma, 50 μ;L; Serum, 50 μ;L; Supernatant, 100 μ;L | |
MCP-5, Scya12 | |
1 hr. 20 min. |