missing translation for 'onlineSavingsMsg'
Learn More

Lovibond™ MD 100 2-in-1 Photometer: Chlorine and pH

Product Code. p-7070893
Click to view available options
Type:
MD 100 2-in-1: Cl + pH (Pwdr)
MD 100 2-in-1: Cl + pH (Tblt)
Unit Size:
Each
This item is not returnable. View return policy

Product Code. 15217539

Brand: Lovibond™ 278030

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Includes

Instrument in a carrying case, three round glass vials with lids, one stirring rod and brush, Vario chlorine free DPD/F10 powder 10mL (100 pack), Vario chlorine total-DPD/F10 powder 10mL(100 pack), Phenol red tablets (100 pack), 4 x AAA batteries, warranty information, Certificate of compliance, instruction manual

Provides accurate and reproducible results very quickly. Lovibond MD 100 Photometer uses high quality interference filters with long-life LEDs as a light source without any moving parts in a transparent sample chamber. Other features include ease of operation, ergonomic design, compact dimensions and safe handling.

The calibration and software-based adjustment options mean that the MD 100 is also suitable for use as a testing instrument. The tests are conducted using either Lovibond tablet reagents with long term stability and a guaranteed minimum five or 10 year shelf life, VARIO powder reagents or using liquid reagents.

  • Waterproof (as defined in IP68, 1 hour at 0.1m)
  • Automatic switch-off
  • Real-time clock and date calibration mode
  • Backlit LCD display (on keypress)
  • Factory calibration and user calibration; Reset to factory calibration possible
  • Internal ring memory for 16 data sets
  • Infrared interface for test data transfer

Reagents Tablets
Each tablet is hermetically sealed within an individual aluminum foil pocket, protecting against challenging environmental conditions. This packaging keeps each tablet in perfect condition, right up until the time it is needed by the user.

Specifications

  • Light source: LED, interference filter (IF) and photosensor in transparent cell chamber
  • Wavelength Accuracy: ±1nm
  • Photometric Accuracy: 3% FS (T = 20°C to 25°C)
  • Photometric Resolution: 0.01 A
  • Temperature: 5° to 40°C
  • Relative Humidity: 30 to 90% (non condensing)
  • Power Supply: 4 AAA batteries
  • Operating Time: approx. 17 hours or 5000 test measurements in continuous mode when display backlight is off
  • Dimensions (L x W x H): 15.5 x 7.5 x 3.5cm
  • Weight: basic unit approx. 260g

Measuring Range

Cat. No. 15267529 (Tablets):Chlorine: 0.01 to 6.0mg/L Cl2/0.1 to 10mg/L Cl2; pH: 6.5 to 8.4

Cat. No. 15267529 (Powder):Chlorine: 0.02 to 2.0mg/L Cl2/0.1 to 8.0mg/L Cl2; pH: 6.5 to 8.4

Note
Specifications for each type of tablet as well as a Certificate of Analysis for each lot is available in the download area at www.lovibond.com.

Specifications

Type MD 100 2-in-1: Cl + pH (Pwdr)
Display Type Backlit LCD
Accuracy 3% FS
Height (Metric) 3.5 cm
Includes Instrument in a carrying case, three round glass vials with lids, one stirring rod and brush, Vario chlorine free DPD/F10 powder 10mL (100 pack), Vario chlorine total-DPD/F10 powder 10mL(100 pack), Phenol red tablets (100 pack), 4 x AAA batteries, warranty information, Certificate of compliance, instruction manual
Length (Metric) 15.5 cm
Memory Internal ring memory for 16 data sets
Light Source LED
Range Chlorine: 0.02 to 2.0mg/L Cl2/0.1 to 8.0mg/L Cl2; pH: 6.5 to 8.4
Resolution 0.01 A
Weight (Metric) 260 g
Width (Metric) 7.5 cm
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.