Learn More
Lovibond™ MD100 3-in-1 photometer kit
MD100 3-in-1 photometer kit for liquid reagents for chlorine 0.02-4.0mg/L pH 6.5-8.4 and total alkalinity 5-200 with tablets includes 3 vials reagents and carry case
Brand:  Lovibond™ 278065
Product Code. 15446151
  • £590.00 / 1 unit
Explore more special offers
Description
Description
The Lovibond™ MD 100 3in1 Pool Photometer is a multi-parameter instrument which allows for fast and easy measurement of pool water quality. Easily test for Chlorine, pH and Total Alkalinity with a combination of Tablet and Liquid reagents.Ideal for poolside testing, the MD 100 comes ready to use with all of the reagents and accessories required for testing.
The MD 100 has several key features that make it a favorite among users.
Waterproof Design
The IP 68 design of this instrument ensures that the MD 100 remains water tight for up to 1 hour at 0.1 meter.
Don't worry about dropping the instrument into a body of water - it floats!
Accurate, Reproducible Results
The optical system of the MD 100 utilizes LEDs and interference filters
which quickly gives you results that you can be confident in.
User Calibration Mode
A one-point calibration is possible.
Additionally, all MD 100 instruments have the ability to display readings directly in ABS or %T units.
One-time-Zero Function
Don't waste time waiting for your instrument to Zero after each test.
When testing a new sample, zero the instrument once and all subsequent tests of that sample do not require you to re-zero the instrument.
Automatic Data Storage with Transfer Capabilities
The MD 100 automatically stores the last 16 readings with time and date stamp.
These results can be transferred to a spreadsheet using the IRiM accessory.
Ergonomic Design
Whether you're using the MD 100 out in the field or in the lab, the design of the instrument makes it easy to handle and view the display.
If you're using the instrument in the lab, the rubber feet on the bottom of the housing eliminates the need for two hand operation or to chase your instrument around the workbench.
Factory Programmed
All MD 100 instruments are supplied ready to use with up to 14 calibration curves.
All you need to do is select the test you need, prepare your sample and test!
Scroll Driven Menu System
The simple interface allows you to quickly reach the test you need.
When the instrument is turned on, it will automatically display the last test used.
2-Year Warranty
Specifications
Specifications
Photometer
LCD
Chlorine, Alkalinity and pH Measurement