Learn More
Lovibond™ MD200 3-in-1 photometer kit for chlorine pH and M-alkalinity with tablet reagents
includes tablet reagents, 3 round vials with lids, carry case, batteries and instruction manual
Brand: Lovibond™ 2889002
Code : NEW
Additional Details : Weight : 1.26800kg
Description
Versatile, compact and: The benchtop photometer MD 200 combines high-quality interference filters with long-term stable LEDs as light sources in a high-precision optic. The result: precise measurement results at all times - no matter where you use it.
Versatile use
The MD 200 photometer can be used in almost all areas of water analysis.
Precise and low maintenance
The high-precision optics, which in addition to long-term stable LEDs as light sources, also offer high-quality interference filters, ensure precise and reproducible measurement results. In addition, these components make the photometer low-maintenance.
User friendly
The devices impress with their ease of use, ergonomic design and compact dimensions, which makes handling the devices easy and safe.
Reagents
Use long-term stable Lovibond™ reagent tablets (5-10 years guaranteed minimum durability) or Lovibond™ liquid reagents.
Specifications
Photometer | |
LCD | |
Chlorine, pH and M-Alkalinity Measurement |