missing translation for 'onlineSavingsMsg'
Learn More

Lovibond™ MD200 COD set-up with reagents

Product Code. 12661735
Click to view available options
Type:
Photometer
Unit Size:
Each
This item is not returnable. View return policy

Product Code. 12661735

Brand: Lovibond™ 2892602

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MD200 COD set-up with reagents includes MD200 COD photometer RD125 reactor tube stand and reagents

The Lovibond™ COD Setups allow highly sensitive and precise water testing with minimum effort. They measure the ST-COD concentration by photometric detection employing a linear relationship between absorbance and concentration.After adding the sample to a Lovibond™ COD tube test (LR, MR according to ISO 15705:2002), it is heated in the reactor for two hours at 150 °C and then analysed in the photometer.The COD Setups comprise the photometer, 25 tube tests for each of the two lower measuring ranges, a reactor for sample digestion and a vial stand.

The Lovibond™ COD Setups allow highly sensitive and precise water testing with minimum effort. They measure the ST-COD concentration by photometric detection employing a linear relationship between absorbance and concentration.

After adding the sample to a Lovibond™ COD tube test (LR, MR according to ISO 15705:2002), it is heated in the reactor for two hours at 150 °C and then analysed in the photometer.

The COD Setups comprise the photometer, 25 tube tests for each of the two lower measuring ranges, a reactor for sample digestion and a vial stand.

Specifications

Type Photometer
Display Type LCD
For Use With (Application) COD
compliance-icons compliance-icons compliance-icons compliance-icons compliance-icons
Product Identifier
  • MD200 Cod vario photometer, complete
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • Acute toxicity Category 4
  • LONG-TERM AQUATIC HAZARD Chronic 1
  • ACUTE AQUATIC Acute 1
  • Carcinogenicity Category 1B
  • Serious eye damage/eye irritation Category 1
  • Substances and mixtures corrosive to metals Category 1
  • Germ cell mutagenicity Category 1B
  • Skin corrosion/irritation Category 1 A
  • Specific target organ toxicity after repeated exposure Category 2
Hazard Statement
  • H290-May be corrosive to metals.
  • H302-Harmful if swallowed.
  • H311-Toxic in contact with skin.
  • H314-Causes severe skin burns and eye damage.
  • H318-Causes serious eye damage.
  • H332-Harmful if inhaled.
  • H340-May cause genetic defects.
  • H350-May cause cancer.
  • H373-May cause damage to organs.
  • H400-Very toxic to aquatic life.
  • H410-Very toxic to aquatic life with long lasting effects.
  • EUH208-Contains potassium dichromate. May produce an allergic reaction.
Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contains : sulphuric acid, mercury sulphate, disilver sulphate, potassium dichromate
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.