missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MDC Monoclonal Antibody (57226), Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen MA523754
This item is not returnable.
View return policy
Description
In ELISAs, this antibody does notcross-react with recombinant mouse (rm) CCL17, rhCCL17, or rhCCL21. Reconstitute at 0.5 mg/mL in sterile PBS. Endoxin level is <0.10 EU per 1 μg of the antibody by the LAL method.
MDC Monoclonal antibody specifically detects MDC in Human samples. It is validated for ELISA, Neutralization, Western Blot.Specifications
MDC | |
Monoclonal | |
0.5 mg/mL | |
PBS with 5% trehalose and No Preservative | |
O00626 | |
CCL22 | |
E. coli-derived recombinant human CCL22/MDC Gly25-Gln93 | |
500 μg | |
Primary | |
Human | |
Antibody | |
IgG2b |
ELISA, Neutralization, Western Blot | |
57226 | |
Unconjugated | |
CCL22 | |
A-152E5.1; Abcd1; ABCD-1; Activated B and dendritic cell-derived; AW060611; CC chemokine ABCD-1; CC chemokine STCP-1; C-C motif chemokine 22; C-C motif chemokine ligand 22; Ccl22; chemokine (C-C motif) ligand 22; DC/B-CK; DCBCK; dendritic cell and B cell derived chemokine; macrophage-derived chemokine; MDC; MDC(1-69); MDC(3-69); MDC(5-69); MDC(7-69); MGC108943; MGC34554; RP24-69A20.1; Scya22; small inducible cytokine A22; SMALL INDUCIBLE CYTOKINE A22 PRECURSOR (CC CHEMOKINE ABCD-1) (ACTIVATED B AND DENDRITIC CELL-DERIVED); small inducible cytokine subfamily A (Cys-Cys) member 22; small inducible cytokine subfamily A (Cys-Cys), member 22; small inducible cytokine subfamily A, member 22; small inducible cytokine subfamily A22; small-inducible cytokine A22; STCP-1; stimulated T cell chemotactic protein 1; stimulated T-cell chemotactic protein 1 | |
Mouse | |
Protein A/G | |
RUO | |
6367 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Lyophilized |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction