Learn More
Invitrogen™ MDK Human ELISA Development Kit (ABTS), PeproTech®, PeproTech®
ELISA
Brand: Invitrogen™ 900-M190
This item is not returnable.
View return policy
Description
Midkine is a retinoic acid-responsive, heparin-binding growth factor expressed in various cell types during embryogenesis. It promotes angiogenesis, cell growth, and cell migration. Midkine is also expressed in several carcinomas, suggesting that it may play a role in tumorigenesis, perhaps through its effects on angiogenesis.Specifications
P21741 | |
4192 | |
2 x 96 Tests | |
Refer to product documentation for component specific storage temperature. |
ELISA | |
Mdk | |
RUO | |
Human |