missing translation for 'onlineSavingsMsg'
Learn More

Thermographic Measurements Ltd Thermax Multi-Change Temperature Indicating Paint

Multi-change timed temperature indicating paint

Brand:  Thermographic Measurements Ltd IPMC135THE250ML

 View more versions of this product

Product Code. 11741199

  • £278.00 / Each
Estimated Shipment: 10-02-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Timed Temperature Paint
  • Solvent-based
  • No primer necessary on most surfaces
  • Colorfast in direct sunlight
  • Fast reaction time
  • Superior performance in normal and hostile environments
  • Ideal for thermal surveys
  • Strong clear color contrasts
  • Resistant to oil, steam and water
  • Requires no additives
  • Specifications

    Specifications

    Timed Temperature Paint (135°C)
    ISO 9001
    250 mL
    Color changing paint to show heat exposure
    Pink
    Insulated vessels, motor components, thermal mapping of equipment
    Product Suggestions

    Product Suggestions

    Videos
    SDS
    Documents

    Documents

    Certificates
    Special Offers

    Special Offers

    Product Content Correction

    Your input is important to us. Please complete this form to provide feedback related to the content on this product.

    Product Title

    By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

    Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.