missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mecamylamine hydrochloride, Tocris Bioscience™
Click to view available options
Quantity:
10 mg
50 mg
Description
Non-competitive nicotinic acetylcholine receptor antagonist. Displays antidepressant-like effects in mice.
Specifications
Specifications
Quantity | 10 mg |
Formula Weight | 203.75 |
Percent Purity | >98% |
Chemical Name or Material | (1 R,2 S,4 S)-rel-N,2,3,3-Tetramethylbicyclo[2.2.1 ]heptan-2-amine hydrochloride |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'