Learn More
Karl Hecht™ Mechanical Manual Counter
A four-figure counter with button for manual counting.
Brand:  Karl Hecht™ 40344010
Product Code. 10338911
  • £27.30 / Each
Estimated Shipment: 07-05-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Counter is held with the forefinger at the finger bow
  • Counter is turned back to zero by using the lateral reset button
  • Counts from 0 to 0000
  • Casing made of chrome-plated steel
  • Ergonomic design; lies comfortably in hand
Specifications
Specifications
Manual Counter
0 to 9999
Chrome-plated steel casing
Each
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Karl Hecht™ Mechanical Manual Counter > Manual counter; 0 to 9999