missing translation for 'onlineSavingsMsg'
Learn More

AMPri™ Med-Comfort Premium Grip Latex Examination Gloves, PPE Cat. III

Product Code. 16826081
Click to view available options
Size:
Large
Medium
Small
X-Large
This item is not returnable. View return policy
You may only order up to the available stock. Where an alternative item has been identified it will be shown below
This item is not returnable. View return policy
You may only order up to the available stock. Where an alternative item has been identified it will be shown below

Non-sterile AMPri™ Med-Comfort latex examination gloves are powder-free and have a textured finish for good grip. Ideal for general laboratory use as well as clinical, pharmaceutical and food use applications, they offer excellent protection against microorganisms (viral, bacteria and fungi).

  • AQL 1.5
  • Premium grip examination gloves
  • Applications include laboratory, clinical, pharmaceutical, dental, and food industry use
  • Protect against chemicals and microorganisms
  • Ambidextrous fit
  • Powder-free
  • Non-sterile
  • Color: white
  • Textured finish
  • Disposable

Specifications

Certifications/Compliance AQL 1.5
Certifications/Compliance (Europe) EN 455-1, EN374-1/4/5, EN420, Food grade
Color White
Disposable Single-use
Laboratory/Cleanroom Use Laboratory
Latex Free No
Material Latex
Size Small
Thickness (Metric) 0.08 mm (Cuff)
Length (Metric) 240 mm
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.