missing translation for 'onlineSavingsMsg'
Learn More

Macherey-Nagel™ Medi-Test Combi 3A™

Perform reliable urinalysis at the point of care. Macherey-Nagel™ Medi-Test Combi 3A™ is a simple Dip & Read urine test strip that provides important patient information in under a minute to simplify the decision making process about further diagnostic and theraputic steps.

Brand:  Macherey-Nagel™ 93030

Product Code. 11310425

  • £14.25 / Pack of 100

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

The use of urine test strips is acknowledged as modern screening method in medical practice.

  • Applications include screenings for detection and control of diabetes and kidney or urinary tract disease
  • Multiparameters dipstick detects Glucose, Ascorbic acid, Protein and pH
  • CE and FDA marked for professional use
  • Presented in pack of 100 strips
Specifications

Specifications

CE, FDA 510(k): K991927
Store below 30°C in dry place, in closed original container
Waived (simple)
Yes
Tube with color guide
Protein: tetrabromophenol blue-protein error indicator, Glucose: glucoseoxidase-peroxidase-chromogen, pH: methyl red / bromothymol blue indicator, Ascorbic acid: Tillmans reagent indicator
100 Pack
Protein: yellow to green-blue, Ascorbic acid: blue to red, Glucose: yellow to green to blue, pH: orange to green to turquois
Yes
Glucose, Ascorbic Acid, Protein, pH
Rapid determination of protein, ascorbic acid, glucose, pH-value in urine for detection/monitoring of diabetes, kidney and urinary tract disease
Cellulose Strip
Plastic Tube
30 to 60 sec.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers