Learn More
Medical Viscosity Standard, 2.0 mPa.s (25.00°C ), Paragon Scientific™
Tested in strict accordance with ASTM D2162. These medical grade viscosity standards are fully traceable to national standards and are also test equipment compatible. Typically used, but not limited to, the calibration and verification of viscosity measuring equipment.
Brand:  Paragon Scientific MGVS20-500
 View more versions of this product
Product Code. 16060230
  • £180.00 / 500mL
Explore more special offers
Description
Description
  • Fully traceable to international standards
  • Certified in accordance to ISO 17025 & ISO 17034 under Paragon's UKAS accreditation
  • Certified in strict accordance with ASTM D2162
  • Tamper evident security packaging
  • 12 month shelf life
  • Manufactured in the United Kingdom
  • Contains antibacterial agent
Specifications
Specifications
500 mL
ISO 17025 / ISO 17034
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Nominal values quoted. Certified Values will vary from batch to batch.