missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Liebherr™ Spark-Free interior, Lab Fridge
For secure, accurate and long-lasting, spark-free interior, fridge storage of non temperature-sensitive products, the Liebherr™ Spark-Free Interior, Lab Fridge is designed for general purpose applications with Liebherr build-quality you can rely on.
Brand: Liebherr™ COOLSAFE 16460
This item is not returnable.
View return policy
Description
- Spark-Free Lab Fridge for storage of solvents and reagents, reach-in, 1-section, width 600mm.
- Temperature range +1 / +15°C, white exterior, commercial-grade moulded polystyrol interior, fan-assisted refrigeration.
- Climate class 4, refrigerant R 600a, connection rating 1,0A, 230v/50/1-ph,energy consumption 328 kWh/a
- This model is intended for the spark-free storage of chemicals where a high level of temperature stability is not required
- ATEX-certified spark-free interior
- Analogue controls with digital temperature display in the door.
- Door lock, reversible hinges
- 40 kg shelf load
Specifications
Laboratory Refrigerator | |
1°C to 15°C | |
Gradient 4.8°C | |
Solid | |
Right, reversible | |
5 | |
Steel | |
40 mm HDPE | |
ATEX Certified, EU Directive 94/9/EC (ATEX 95), ISO 14001, ISO 9001 | |
Digital, External | |
White | |
Automatic | |
61 cm | |
164 cm | |
60 cm | |
1.5 A | |
220 - 240 V 50 Hz 1.5 A | |
346 kWh/a | |
59 kg |
333 L | |
1.0°C | |
R 600a | |
1 | |
0 | |
40 kg | |
White-coated Steel | |
Polystyrol | |
Tall | |
No | |
Analog | |
Forced-air | |
44 cm | |
145.2 cm | |
47 cm | |
220/240 V | |
50 Hz | |
64 kg |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Liebherr™ Spark-Free interior, Lab Fridge > Capacity: 333 L; Cabinet Type: Tall
Spot an opportunity for improvement?Share a Content Correction