Learn More
Liebherr™ Pharmacy Fridge
For secure, accurate and long-lasting fridge storage of temperature-sensitive pharmacy products, the Liebherr™ Pharmacy Fridge is fully-equipped for pharmacy applications with Liebherr build-quality you can rely on. DIN 58345 compliant.
Brand: Liebherr™ MKv 3910
Code : NEW
Additional Details : Weight : 75.00000kg Transport : UN number : 2800 Chem class : 8
Description
- DIN 58345 compliant, width 600mm.
- Temperature set to +5°C, white exterior, commercial-grade moulded polystyrol interior, fan-assisted refrigeration.
- Climate class 4, refrigerant R 600a.
- Digital controller with 0,1°C resolution, password keypad lock, RS 485 interface and volt-free alarm contact
- Access port for external temperature probe, additional safety thermostat
- Alarm log of last 3 events with min/max temperatures, duration etc.
- Integrated battery for continuous alarm and monitoring for min. 12 hrs in case of power failure
- Door lock, reversible hinges, self-closing door
- 45 kg shelf load
Specifications
Pharmacy refrigerator | |
5°C | |
Pt100 | |
R 600a | |
Steel | |
5 | |
White-coated Steel | |
DIN 58345 | |
No | |
Digital | |
SmartCoolingHub | |
184 H x 60 W x 61.5 D cm | |
43.5 cm | |
163.5 cm | |
44 cm | |
220/240 V | |
50 Hz | |
74 kg |
361 L | |
0.1°C | |
Gradient 4.1°C | |
Solid | |
0 | |
45 kg | |
White Plastic Containers | |
Tall | |
White | |
Automatic | |
RS 485 | |
61.5 cm | |
184 cm | |
60 cm | |
1.5 A | |
220 - 240 V 50 Hz | |
309 kWh/a | |
68 kg |