Learn More
Liebherr™ Pharmacy Fridge
For secure, accurate and long-lasting fridge storage of temperature-sensitive pharmacy products, the Liebherr™ Pharmacy Fridge is fully-equipped for pharmacy applications with Liebherr build-quality you can rely on. DIN 58345 compliant.
Brand: Liebherr™ MKUv 1613
1395.16 GBP valid until 2024-08-31
Use promo code "21622" to get your promotional price.
Code : NEW
Additional Details : Weight : 45.00000kg Transport : UN number : 2794 Chem class : 8
Description
- DIN 58345 compliant, width 600mm.
- Temperature set to +5°C, white exterior, commercial-grade moulded polystyrol interior, fan-assisted refrigeration.
- Climate class 4, refrigerant R 600a.
- Digital controller with 0,1°C resolution, password keypad lock, RS 485 interface and volt-free alarm contact
- Access port for external temperature probe, additional safety thermostat
- Alarm log of last 3 events with min/max temperatures, duration etc.
- Integrated battery for continuous alarm and monitoring for min. 12 hrs in case of power failure
- Door lock, reversible hinges, self-closing door
- 45 kg shelf load
Specifications
Pharmacy refrigerator | |
5°C | |
Pt100 | |
R 600a | |
Steel | |
45 kg | |
White Plastic Containers | |
Underbench | |
No | |
Digital | |
LED internal light | |
RS 485 | |
61.5 cm | |
82 cm | |
59.7 cm | |
1.0 A | |
220 - 240 V 50 Hz | |
369 kWh/a | |
45 kg |
152 L | |
0.1°C | |
Gradient 5.1°C | |
Glass | |
3 | |
White-coated Steel | |
DIN 58345 | |
LED | |
White | |
Automatic | |
SmartCoolingHub | |
82 H x 60 W x 61.5 D cm | |
43.5 cm | |
67 cm | |
44 cm | |
220/240 V | |
50 Hz | |
48 kg |