missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PAL International™ Medipal™ 3 in 1 Disinfectant Wipes
2.0 L canister with 240 disinfectant wipes
Brand: PAL International™ W520110MPCE
13.02 GBP valid until 2024-12-31
Use promo code "21676" to get your promotional price.
This item is not returnable.
View return policy
Description
- Developed in response to a growing clinical need for a single cleaning & disinfectant wipe that is effective against a wider range of pathogens
- Wipes are alcohol free and intended for cleaning and disinfecting non porous hard surfaces, equipment and non invasive medical devices within healthcare environments
- Proven to kill most micro organisms such as bacteria, viruses & yeasts
- The wipes have been independently tested to a range of European standards (EN) including thenew European test EN16615:2015 (4-field test)
- Alcohol & PHMB Free
- 1 minute contact time
- Suitable for storing on trolleys, countertops or wall mounted in a dispenser bracket
Specifications
Cleans and Disinfects | |
240 wipes in a 2.0 L canister |
Safety and Handling
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
Supplemental information
- medipal disinfectant wipes
Signal Word
- Warning
Hazard Category
- LONG-TERM AQUATIC HAZARD Chronic 3
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 2
Hazard Statement
- H412-Harmful to aquatic life with long lasting effects.
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- EUH208-Contains polyhexamethylene biguanide hydrochloride. May produce an allergic reaction.
Precautionary Statement
- P273-Avoid release to the environment.
Supplemental information
- MIXTURE LIST-Contains : alkyl C12-16 dimethyl benzyl ammonium chloride, poly (hexamethylene biguanide) hydrochloride, 5-chlo
Correction du contenu d'un produit
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.
Nom du produit
Vous avez repéré une opportunité d'amélioration ?Partager une correction de contenu