Learn More
PAL International™ Medipal™ Medical Disinfectant Wipes
Supports healthcare institutions to achieve both their clinical and budgetary targets. Using Medipal™ wipes will help infection prevention teams to fight infection and ensure healthy, clinically clean environments for patients.
Brand:  PAL International™ S522110MPCE
11.44 GBP valid until 2024-12-31
Use promo code "21676" to get your promotional price.
Product Code. 15824165
  • £15.00 / Pack of 200
In Stock
to see stock.
Edge
Explore more special offers
Add to basket
Description
Description
  • They contain a powerful dual biocidal solution, highly effective from 30 seconds against a wide range of pathogenic microorganisms and are intended for cleaning and disinfecting surfaces of non invasive medical devices
  • Medipal™ Disinfectant Wipes are bactericidal (including TB), virucidal, yeasticidal and fungicidal
  • They are tested to a range of EN standards including EN13727, EN14476, EN14348, EN14561, EN14563
  • They are alcohol free and gentle on surfaces, making them suitable for everyday use
  • They can be used in combination with Medipal High Level Disinfectant Wipes in areas of particular high risk
  • Low foaming & low residue
Specifications
Specifications
Didecyldimethylammonium chloride (DDAC) & Polyhexamethylene biguanide hydrochloride (PHMB)
Soft Pack
Easy to transport and dispense, these wipes are suitable for general everyday cleaning and disinfection.
200 Wipes/Pk.
Product Suggestions
Product Suggestions
Videos
Safety and Handling
Safety and Handling
Product Identifier
  • Medipal 3in1 Disinfectant Wipes
Signal Word
  • Warning
Hazard Category
  • LONG-TERM AQUATIC HAZARD Chronic 3
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H412-Harmful to aquatic life with long lasting effects.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Supplemental information
  • MIXTURE LIST-Contains : 2-aminoethanol, didecyldimethylammoniumchloride, N-(3-aminopropyl)-N-dodecylpropane-1,3-diamine, al