missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ Medium 199, Hanks' Balanced Salts, powder

Product Code. p-7076084
Click to view available options
Quantity:
10 L
1 L
Unit Size:
1 L
10 L
This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 

Product Code. 10165142

Brand: Gibco™ 10012011

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 

Medium 199, Hanks' Balanced Salts, powder

Medium 199 was originally developed for nutritional studies of chick embryo fibroblasts. It has broad species applicability, particularly for cultivation of non-transformed cells. Medium 199 is widely used in virology, vaccine production, and in vitro cultivation of primary explants of mouse pancreatic epithelium, and rat lens tissues. We offer a variety of Gibco™ Medium 199 modifications for a range of cell culture applications. Find the right formulation using the media selector tool.

Compared to other basal media, Medium 199 contains unique components, including adenine, adenosine, hypoxanthine, thymine, and additional vitamins. This Medium 199 formulation contains Hanks' salts for use without CO2.

Product Intended Use
For in vitro diagnostic use. CAUTION: Not for human or animal therapeutic use. Uses other than the intended use may be a violation of local law.

cGMP Manufacturing
Gibco™ Medium 199 is manufactured at a cGMP compliant facility located in Paisley, Scotland, UK. The facility is registered with the FDA as a medical device manufacturer and is certified to the ISO 13485 standard.

Medium 199 contains no proteins, lipids, or growth factors. Therefore, MEM requires supplementation, commonly with 10% Fetal Bovine Serum (FBS). This Medium 199 formulation does not require a CO2 environment to maintain physiological pH.

Powder forms of Gibco™ cell culture medium require sodium bicarbonate supplementation, pH adjustment, and filtration at the time of preparation (see protocol for details).
TRUSTED_SUSTAINABILITY

Specifications

Form Powder
Product Type Medium 199 Hanks' Balanced Salts
With Additives Hanks’ Balanced Salt solution (HBSS), Glutamine, HEPES, Phenol Red
Without Additives No Sodium Bicarbonate
For Use With (Application) For cell culture, mammalian cell culture, vaccine production and In-vitro cultivation
Quantity 1 L
Certifications/Compliance ISO 13485 standards, FDA
pH 7 to 7.4
Content And Storage 2 to 8°C
Description Hanks' Balanced Salts, Medium 199, 1L, Powdered Form, L-Glutamine, HEPES Buffer, 2 to 8°C Storage Temperature, 36 Months Shelf Life, For Mammalian Cell Culture, Vaccine Production and In-vitro Cultivation
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.