missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Gibco™ Medium 199, HEPES
Medium 199, HEPES
£17.52 - £156.00
Specifications
Certifications/Compliance | ISO 13485 standards, FDA |
---|---|
Content And Storage | 2 to 8°C |
Description | Medium 199, 500mL, 1 X Concentration, Liquid Form, With L-glutamine, Phenol Red, HEPES, Sodium Pyruvate, 7.1 to 7.3 pH, 12 Months Shelf Life, 270 to 330mOsm/kg Osmolality, 2 to 8°C Storage Conditions, Used for Mammalian Cell Culture |
Concentration | 1 X |
Form | Liquid |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11570596
|
Gibco™
22340020 |
500 mL |
£17.52
500mL |
In Stock Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
11580596
|
Gibco™
22340087 |
500 mL |
£156.00
Pack of 10 |
In Stock Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
Medium 199 was originally developed for nutritional studies of chick embryo fibroblasts. It has broad species applicability, particularly for cultivation of non-transformed cells. Medium 199 is widely used in virology, vaccine production, and in vitro cultivation of primary explants of mouse pancreatic epithelium, and rat lens tissues. We offer a variety of Gibco™ Medium 199 modifications for a range of cell culture applications. Find the right formulation using the media selector tool.Compared to other basal media, Medium 199 contains unique components, including adenine, adenosine, hypoxanthine, thymine, and additional vitamins. Medium 199 is available with Earle's salts for use in a CO2 incubator, or with Hanks' salts for use without CO2.
Product Intended Use
For in vitro diagnostic use. CAUTION: Not for human or animal therapeutic use. Uses other than the intended use may be a violation of local law.
cGMP Manufacturing and Quality System
Gibco™ Medium 199 is manufactured at a cGMP compliant facility, located in Paisley, Scotland, UK. The facility is registered with the FDA as a medical device manufacturer and is certified to the ISO 13485 standard. For supply chain continuity, we offer a comparable Gibco™ Medium 199 product made in our Grand Island facility. This facility is registered with the FDA as a medical device manufacturer and is certified to the ISO 13485 standards.
Medium 199 contains no proteins, lipids, or growth factors. Medium 199 uses a sodium bicarbonate buffer system (2.2 g/L) and therefore requires a 5–10% CO2 environment to maintain physiological pH.
Specifications
ISO 13485 standards, FDA | |
Medium 199, 500mL, 1 X Concentration, Liquid Form, With L-glutamine, Phenol Red, HEPES, Sodium Pyruvate, 7.1 to 7.3 pH, 12 Months Shelf Life, 270 to 330mOsm/kg Osmolality, 2 to 8°C Storage Conditions, Used for Mammalian Cell Culture | |
Liquid | |
Glutamine, HEPES, Phenol Red, Sodium Pyruvate | |
7.1 to 7.3 | |
For in-vitro diagnostic use |
2 to 8°C | |
1 X | |
Medium 199 | |
Mammalian cell culture | |
500 mL |