missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Gibco™ Medium 199, low sodium bicarbonate

Medium 199, low sodium bicarbonate
Brand: Gibco™ 31153026
This item is not returnable.
View return policy
Description
Medium 199 was originally developed for nutritional studies of chick embryo fibroblasts. It has broad species applicability, particularly for cultivation of non-transformed cells. Medium 199 is widely used in virology, vaccine production, and in vitro cultivation of primary explants of mouse pancreatic epithelium, and rat lens tissues. We offer a variety of Gibco™ Medium 199 modifications for a range of cell culture applications. Find the right formulation using the media selector tool.Compared to other basal media, Medium 199 contains unique components, including adenine, adenosine, hypoxanthine, thymine, and additional vitamins. Medium 199 is available with Earle's salts for use in a CO2 incubator, or with Hanks' salts for use without CO2.
Product Use
For Research Use Only: Not intended for animal or human diagnostic or therapeutic use.
cGMP Manufacturing and Quality System
Gibco™ Medium 199 is manufactured at a cGMP compliant facility located in Paisley, Scotland, UK. The facility is registered with the FDA as a medical device manufacturer and is certified to the ISO 13485 standard.
Medium 199 contains no proteins, lipids, or growth factors. This Medium 199 uses a sodium bicarbonate buffer system (1.25 g/L) and therefore requires a 3-5% CO2 environment to maintain physiological pH.

Specifications
ISO 13485 standards, FDA | |
Medium 199, 500mL, 1 X Concentration, Liquid Form, With L-glutamine, Phenol Red, Sodium Bicarbonate, Sodium Pyruvate, 7 to 7.2 pH, 12 Months Shelf Life, 2 to 8°C Storage Conditions, Soluble in Water, Used for Mammalian Cell Culture | |
Liquid | |
Glutamine, Phenol Red, Sodium Bicarbonate, Sodium Pyruvate | |
7 to 7.2 | |
For research use only |
2 to 8°C | |
1 X | |
Medium 199 | |
Mammalian cell culture | |
500 mL |
Product Suggestions
Customers who viewed this item also viewed.
Viewing 1-5 of 10
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Gibco™ Medium 199, low sodium bicarbonate > 500mL
Spot an opportunity for improvement?Share a Content Correction