missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Gibco™ Human Large Vessel Endothelial Cell Basal Medium (formerly Medium 200), Phenol Red Free
Medium 200PRF
Brand: Gibco™ M200PRF500
Code : 50
Additional Details : Weight : 0.50000kg
Description
Human Large Vessel Endothelial Cell Basal Medium (formerly “Medium 200”) is a sterile, liquid tissue culture medium intended for use as one component in a complete culture environment for normal human large vessel endothelial cells. When supplemented with low serum growth supplement (LSGS), this medium supports the plating and proliferation of endothelial cells at culture densities between 2.5 x 103 cells/cm2 and 1 x 105 cells/cm2. Additional applications may include primary isolation of endothelial cells. This medium is also available with phenol red (Cat. No. M200500).Human Large Vessel Endothelial Cell Basal Medium is a basal culture medium containing essential and non-essential amino acids, vitamins, other organic compounds, trace minerals, and inorganic salts. This medium does not contain antibiotics, antimycotics, hormones, growth factors, or proteins. It is HEPES and bicarbonate buffered and designed for use in an incubator with an atmosphere of 5% CO2/95% air. To support the plating and proliferation of human large vessel endothelial cells, this medium must be supplemented with LSGS (Cat. No. S-003-10; kit = Cat. No. S-003-K).
Specifications
4 degrees C | |
Mammalian Cell Culture | |
Basal Medium | |
Sterile-filtered | |
No Phenol Red | |
Cascade Biologics™ |
Endothelial Cells | |
Liquid | |
Sterile-filtered | |
HEPES | |
Sustainable packaging | |
Room Temperature |
For Research Use Only. Not for use in diagnostic procedures.