missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ Medium 254
GREENER_CHOICE

Medium 254

Brand:  Gibco™ M254500

75.76 GBP valid until 2024-12-31
Use promo code "21745" to get your promotional price.


Product Code. 10236533

  • £79.75 / 500mL

Please to purchase this item. Need a web account? Register with us today!

Edge
Explore more special offers

For Research Use Only. All usage must comply with product instructions.
 
This item is not returnable. View return policy

Description

Description

A sterile, liquid medium prepared with 200μM calcium chloride for the culture of human epidermal melanocytes. This basal medium requires the addition of HMGS (S-002-5) or HMGS-2 (S-016-5) prior to use.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Store in refrigerator (2–8°C) and protect from light.
Mammalian Cell Culture
Medium 254
Sterile-filtered
Calcium
500 mL
Melanocytes
Liquid
Sterile-filtered
HEPES, Phenol Red
Cascade Biologics™
Room Temperature
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only. Not for use in diagnostic procedures.