Learn More
Medroxyprogesterone acetate, 97%, Thermo Scientific Chemicals
£40.40 - £96.50
Chemical Identifiers
CAS | 71-58-9 |
---|---|
Molecular Formula | C24H34O4 |
Molecular Weight (g/mol) | 386.52 |
InChI Key | PSGAAPLEWMOORI-PEINSRQWSA-N |
Synonym | medroxyprogesterone acetate, medroxyprogesterone 17-acetate, metigestrona, provera, depo-provera, gestapuran, farlutin, perlutex, veramix, methylacetoxyprogesterone |
PubChem CID | 6279 |
ChEBI | CHEBI:6716 |
IUPAC Name | [(6S,8R,9S,10R,13S,14S,17R)-17-acetyl-6,10,13-trimethyl-3-oxo-2,6,7,8,9,11,12,14,15,16-decahydro-1H-cyclopenta[a]phenanthren-17-yl] acetate |
SMILES | CC1CC2C(CCC3(C2CCC3(C(=O)C)OC(=O)C)C)C4(C1=CC(=O)CC4)C |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15587719
|
Thermo Scientific Acros
461120010 |
1 g |
£40.40
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15597719
|
Thermo Scientific Acros
461120050 |
5 g |
£96.50
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
71-58-9 | |
386.52 | |
medroxyprogesterone acetate, medroxyprogesterone 17-acetate, metigestrona, provera, depo-provera, gestapuran, farlutin, perlutex, veramix, methylacetoxyprogesterone | |
CHEBI:6716 | |
CC1CC2C(CCC3(C2CCC3(C(=O)C)OC(=O)C)C)C4(C1=CC(=O)CC4)C |
C24H34O4 | |
PSGAAPLEWMOORI-PEINSRQWSA-N | |
6279 | |
[(6S,8R,9S,10R,13S,14S,17R)-17-acetyl-6,10,13-trimethyl-3-oxo-2,6,7,8,9,11,12,14,15,16-decahydro-1H-cyclopenta[a]phenanthren-17-yl] acetate |
Specifications
71-58-9 | |
1% max. | |
96% min. (HPLC) | |
C24H34O4 | |
Solubility in water: insoluble. | |
CC1CC2C(CCC3(C2CCC3(C(=O)C)OC(=O)C)C)C4(C1=CC(=O)CC4)C | |
386.52 | |
CHEBI:6716 | |
97% |
206.0°C to 208.0°C | |
Authentic | |
Glass Bottle | |
medroxyprogesterone acetate, medroxyprogesterone 17-acetate, metigestrona, provera, depo-provera, gestapuran, farlutin, perlutex, veramix, methylacetoxyprogesterone | |
PSGAAPLEWMOORI-PEINSRQWSA-N | |
[(6S,8R,9S,10R,13S,14S,17R)-17-acetyl-6,10,13-trimethyl-3-oxo-2,6,7,8,9,11,12,14,15,16-decahydro-1H-cyclopenta[a]phenanthren-17-yl] acetate | |
6279 | |
386.52 | |
Medroxyprogesterone acetate |
Safety and Handling
GHS H Statement
Suspected of causing cancer.
May damage fertility.
May damage the unborn child.
GHS P Statement Obtain special instructions before use. Wear protective gloves/protective clothing/eye protection/face protection. IF exposed or concerned: Get medical advice/attention.
GHS Signal Word: Danger
missing translation for 'einecsNumber' : 200-757-9
- Medroxyprogesterone acetate
Signal Word
- Danger
Hazard Category
- Carcinogenicity Category 2
- Reproductive toxicity Category 1B
Hazard Statement
- H360FD-May damage fertility. May damage the unborn child.
- H351-Suspected of causing cancer.
Precautionary Statement
- P201-Obtain special instructions before use.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P308+P313-IF exposed or concerned: Get medical advice/attention.
RUO – Research Use Only