missing translation for 'onlineSavingsMsg'
Learn More

Mefenamic acid, MedChemExpress

Product Code. p-200099746
Encompass_Preferred
Click to view available options
Quantity:
5 g
10 g
Unit Size:
10g
5g
This item is not returnable. View return policy

Product Code. 16468528

Brand: Medchem Express HYB0574

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Mefenamic acid is a non-steroidal anti-inflammatory agent, acting as a competitive inhibitor of hCOX-1 and hCOX-2, with IC50s of 40 nM and 3 μM for hCOX-1 and hCOX-2, respectively.

Mefenamic acid is a non-steroidal anti-inflammatory agent, acting as a competitive inhibitor of hCOX-1 and hCOX-2, with IC50s of 40 nM and 3 μM for hCOX-1 and hCOX-2, respectively.

Specifications

Chemical Name or Material Mefenamic acid
Physical Form Solid
CAS 61-68-7
Color White
Quantity 10 g
Purity Grade Notes Research
Molecular Formula C15H15NO2
For Use With (Application) COVID-19-immunoregulation
Solubility Information DMSO : ≥ 100 mg/mL (414.44 mM) ∣H2O : < 0.1 mg/mL (insoluble)
SMILES O=C(O)C1=CC=CC=C1NC2=CC=CC(C)=C2C
Molecular Weight (g/mol) 241.29
Formula Weight 241.29
Percent Purity 99.97%
Grade Research
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.