Learn More
STEMCELL Technologies MegaCult™-C Complete Kit with Cytokines
Complete kit including collagen, media, staining kit and chamber slides for human CFU-Mk assays
Brand: STEMCELL Technologies 04971
This item is not returnable.
View return policy
Description
- MegaCult™-C Complete Kit with Cytokines includes all reagents required for the optimal growth of megakaryocyte progenitors (CFU-Mk) within double chamber slides and for the detection of CFU-Mk colonies by immunocytochemical staining
- MegaCult™-C medium is optimized for the growth of CFU-Mk in human bone marrow, mobilized peripheral blood and cord blood samples
- It is suitable for use with CD34+-enriched cells, mononuclear cells and cells isolated by other purification methods
Specifications
Hematopoietic Stem and Progenitor Cells | |
Cell Culture, Colony Assay, Functional Assay | |
1 Kit |