Learn More
STEMCELL Technologies MegaCult™-C Double Chamber Slides
For the culture, fixation and staining of collagen-based cultures
Brand: STEMCELL Technologies 04813
Additional Details : Weight : 0.22220kg
Description
- Double Chamber Slide Kit contains 48 double chamber slides, 48 filter cards, and 48 spacers
- The double chamber slides are recommended for performing colony assays in collagen gels using MegaCult™-C or other collagen-based media, for the detection of hematopoietic progenitor cells
- The chamber slide system allows for culture, fixation, and immunocytochemical staining of hematopoietic colony assays to be performed on the same slide
- Filter cards and spacers are used for the dehydration of collagen-based gels following culture
- The kit provides sufficient slides, filter cards, and spacers to perform 24 duplicate assays (two slides) or 48 single assays (one slide)
Specifications
Cell Culture; Colony Assay | |
16/Box |