missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Uvex™ Megasonic Goggles
14.18 GBP valid until 2024-12-31
Use promo code "23255" to get your promotional price.
Use promo code "23255" to get your promotional price.
Click to view available options
Lens Tint:
Brown (23%)
Clear
Description
The sporty, ergonomic design and ultra-comfortable fit means these goggles both protect and allow you to get on with your day.
General features
- Functional goggles in ski goggle design with panoramic view
- Unique, frameless lens design with maximum optimised field of view for better hazard perception and increased safety
- Robust polycarbonate lens with proven uvex supravision coating technology – permanently anti-fog and scratch-resistant lens
- Sustainable benefit: headband can be replaced if necessary
- Metal-free
Protection features
- Certified in line with EN 166 (personal eye protection) and EN 170 (UV filters)
- UV400 – 100% protection against dangerous UVA and UVB radiation to 400 nm
- Increased mechanical strength – BT marking (120 m/s; tested under extreme temperatures -5°C, +55°C)
Comfort features
- Soft, adaptive face pad provides a universal fit for different head and face shapes and a tight fit without pressure points
- Generous internal volume and recesses for the side arms of the glasses allow comfortable wear without pressure points, even for glasses wearers
- Indirect ventilation system increases air circulation
- 30 mm wide length adjustable fabric headband for optimum hold and individual adjustment
Specifications
Lens Type | Chemical-resistant |
Lens Coating | Anti-fog, Scratch-resistant |
Color | Gray, Black |
Certifications/Compliance (Europe) | EN 166, EN 172 |
Lens Tint | Brown (23%) |
Headband Material | Textile |
Lens Material | Polycarbonate |
UV Protection | UV400 |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'