missing translation for 'onlineSavingsMsg'
Learn More

Megestrol Acetate, Tocris Bioscience™

Product Code. 30000889
Click to view available options
Quantity:
50 mg
This item is not returnable. View return policy
This item is not returnable. View return policy

Synthetic progesterone analog

Synthetic progesterone analog. Dose-dependently inhibits growth of HepG2 cells (IC50 = 260 μM) in vitro and in vivo and reduces tumor volume. High dosages improve weight gain in tumor-bearing nude mice. Orally bioavailable.

Specifications

CAS 595-33-5
Molecular Formula C24H32O4
Linear Formula C24H32O4
Quantity 50 mg
Synonym megestrol acetate, niagestin, megace es, megestryl acetate, megace, megeron, ovaban, ovarid, pallace, magestin
Solubility Information Soluble to 50mM in DMSO and to 20mM in ethanol
InChI Key RQZAXGRLVPAYTJ-GQFGMJRRSA-N
SMILES CC1=CC2C(CCC3(C2CCC3(C(=O)C)OC(=O)C)C)C4(C1=CC(=O)CC4)C
IUPAC Name [(8R,9S,10R,13S,14S,17R)-17-acetyl-6,10,13-trimethyl-3-oxo-2,8,9,11,12,14,15,16-octahydro-1H-cyclopenta[a]phenanthren-17-yl] acetate
Molecular Weight (g/mol) 384.516
PubChem CID 11683
Formula Weight 384.51
Percent Purity >98%
Physical Form Solid
Chemical Name or Material 17-(Acetyloxy)-6-methylpregna-4,6-diene-3,20-dione
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.