missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific Chemicals Meglumine antimoniate
£87.10 - £278.00
Chemical Identifiers
CAS | 133-51-7 |
---|---|
Molecular Formula | C7H17NO5·HO3Sb |
Molecular Weight (g/mol) | 365.97 |
InChI Key | YPAUTKZODZLFQM-OVHUINQISA-K |
PubChem CID | 131664208 |
IUPAC Name | antimony(3+);(2R,3R,4R,5S)-6-(methylamino)hexane-1,2,3,4,5-pentol;trihydroxide;hydrate |
SMILES | CNCC(C(C(C(CO)O)O)O)O.O.[OH-].[OH-].[OH-].[Sb+3] |
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
133-51-7 | |
365.97 | |
131664208 | |
CNCC(C(C(C(CO)O)O)O)O.O.[OH-].[OH-].[OH-].[Sb+3] |
C7H17NO5·HO3Sb | |
YPAUTKZODZLFQM-OVHUINQISA-K | |
antimony(3+);(2R,3R,4R,5S)-6-(methylamino)hexane-1,2,3,4,5-pentol;trihydroxide;hydrate |
Specifications
133-51-7 | |
YPAUTKZODZLFQM-OVHUINQISA-K | |
antimony(3+);(2R,3R,4R,5S)-6-(methylamino)hexane-1,2,3,4,5-pentol;trihydroxide;hydrate | |
131664208 | |
Meglumine antimoniate |
C7H17NO5·HO3Sb | |
CNCC(C(C(C(CO)O)O)O)O.O.[OH-].[OH-].[OH-].[Sb+3] | |
365.97 | |
365.97 |
Safety and Handling
GHS Signal Word: Warning
missing translation for 'einecsNumber' : 205-108-3
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
Supplemental information
- Meglumine antimoniate
Signal Word
- Warning
Hazard Category
- Acute toxicity Category 4
- LONG-TERM AQUATIC HAZARD Chronic 2
Hazard Statement
- H302-Harmful if swallowed.
- H332-Harmful if inhaled.
- H411-Toxic to aquatic life with long lasting effects.
Precautionary Statement
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P264-Wash face, hands and anyexposed skin thoroughly after handling.
- P273-Avoid release to the environment.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P312-Call a POISON CENTER/doctor/if you feel unwell.
Supplemental information
- MIXTURE LIST-Contains : 1-Deoxy-1-(methylamino)-D-glucitol
RUO – Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title