missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MEK6 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen 720309
This item is not returnable.
View return policy
Description
These Polyclonal antibodies are of rabbit origin developed by immunizing animals with proteins or peptides. The polyclonal antibody is purified by affinity purification from the rabbit sera generated after immunizing the rabbits with a specific type of protein or peptide. The purified antibody is tested for its functionality in various relevant research applications. The antibody is developed for Research Use Only and is non-hazardous or non-infectious in nature. This antibody is predicted to react with Monkey, PigHand Horse
MKK6 (MEK6, MAP2K6) is a member of the dual specificity protein kinase family, which functions as a mitogen-activated protein (MAP) kinase kinase. MAP kinases, also known as extracellular signal-regulated kinases (ERKs), act as an integration point for multiple biochemical signals. This protein phosphorylates and activates p38 MAP kinase in response to inflammatory cytokines or environmental stress. As an essential component of p38 MAP kinase mediated signal transduction pathway, this gene is involved in many cellular processes such as stress induced cell cycle arrest, transcription activation and apoptosis. Tissue specificity: Isoform 2 is only expressed in skeletal muscle. Isoform 1, on the other hand, is found in skeletal muscle, heart, and in lesser extent in liver or pancreas.Specifications
MEK6 | |
Polyclonal | |
Unconjugated | |
MAP2K6 | |
Dual specificity mitogen-activated protein kinase kinase 6; MAP kinase kinase 6; MAP2K6; MAPK/ERK kinase 3; MAPK/ERK kinase 6; MAPKK 3; MAPKK 6; MAPKK3; MAPKK6; MEK 3; MEK 6; MEK3; MEK6; mitogen activated protein kinase kinase 6; mitogen-activated protein kinase kinase 6; MKK3; MKK6; PRKMK3; Prkmk6; protein kinase, mitogen activated, kinase 6; protein kinase, mitogen-activated, kinase 6 (MAP kinase kinase 6); SAPK kinase 2; SAPK kinase 3; SAPKK2; SAPKK-2; SAPKK3; SAPKK-3; SKK3; Stress-activated protein kinase kinase 3 | |
Rabbit | |
Affinity chromatography | |
RUO | |
26399, 5608 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Flow Cytometry, Immunocytochemistry, Western Blot | |
0.5 mg/mL | |
PBS with 0.1% BSA, 30% glycerol and 0.09% sodium azide; pH 7.4 | |
P52564, P70236 | |
MAP2K6 | |
Peptides corresponding to Human MAP2K6 (aa 5-23, 317-334) | |
100 μg | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
Research Use Only