missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Fisherbrand™ Meker Burner with Tétine Connection, 24mm Opening
Provides a stable flame and excellent air-gas mixture for superior combustion.
Brand: Fisherbrand™ 4101/22BIOM
Description
- 24mm diameter burner opening
- 160mm overall height
- Gas supply regulation by needle valve
- Nickel plated steel to guarantee a stable and homogeneous flame
- For use with natural gas/methane or butane/propane
Specifications
Meker Burner | |
Consumption of 62L/hr | |
160 mm | |
1 Ea. |
Natural Gas | |
24 mm | |
T-Tine Connector |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Fisherbrand™ Meker Burner with Tétine Connection, 24mm Opening > Height: 160mm
Spot an opportunity for improvement?Share a Content Correction