missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Melamine, 99%, Thermo Scientific Chemicals
Click to view available options
Quantity:
5 g
100 g
1 kg
10 kg
Packaging:
Glass bottle
Plastic bottle
Plastic drum
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications
Specifications
Melting Point | 354°C |
Color | White |
Density | 1.5730g/mL |
Infrared Spectrum | Authentic |
Assay Percent Range | 99% |
Packaging | Plastic bottle |
Quantity | 1 kg |
Beilstein | 26, 245 |
Merck Index | 15, 5880 |
Specific Gravity | 1.573 |
Show More |
Safety and Handling

Product Identifier
- Melamine
- Warning
- Carcinogenicity Category 2
- Reproductive toxicity Category 2
- Specific target organ toxicity after repeated exposure Category 2
- H351-Suspected of causing cancer.
- H361f-Suspected of damaging fertility.
- H373-May cause damage to organs.
- P201-Obtain special instructions before use.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P308+P313-IF exposed or concerned: Get medical advice/attention.
- MIXTURE LIST-Contains : Melamine
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Melamine, 99%, Thermo Scientific Chemicals >
Spot an opportunity for improvement?Share a Content Correction