missing translation for 'onlineSavingsMsg'
Learn More

VITLAB™ Melamine Formaldehyde Instrument Trays

Use this practical tray for instruments, tools and sensitive utensils. VITLAB™ Melamine Formaldehyde Instrument Trays are stable and self-supporting with smooth, easy to clean surfaces.

£60.05 - £207.00

Specifications

Color White
Material Melamine
Type Instrument Tray
For Use With Practical tray for instruments, tools, and sensitive utensils
View More Specs

Products 3
Product Code Brand Dimensions (L x W x H) Price Quantity & Availability  
Product Code Brand Dimensions (L x W x H) Price Quantity & Availability  
15791996
View Documents
VITLAB™
72398
290 x 160 x 60 mm
£122.00
Pack of 5
In Stock
to see stock.
Add to basket
 
15781996
View Documents
VITLAB™
72298
350 x 250 x 40 mm
£207.00
Pack of 5
In Stock
to see stock.
Add to basket
 
15242619
View Documents
VITLAB™
72098
190 x 150 x 40 mm
£60.05
Pack of 5
Estimated Shipment: 31-01-2025
to see stock.
Add to basket
 
Description

Description

  • White instrument tray
  • Tall shape
  • Rounded corners
  • Fitting lid made of PS, please order separately
Specifications

Specifications

White
Melamine
Instrument Tray
Practical tray for instruments, tools, and sensitive utensils
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.