Learn More
Thermo Scientific Chemicals Melanocyte-Stimulating Hormone-Release Inhibiting Factor, synthetic
Inhibits α-melanocyte-stimulating hormone (α-MSH) release. Increases brain dopamine turnover and inhibits physiological and behavioral opioid effects in vivo. Role in vascular homeostasis.
£44.52 - £156.00
Specifications
Molecular Weight (g/mol) | 284.36 |
---|---|
Packaging | Glass bottle |
Form | Lyophilized |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15450117
|
Thermo Scientific Chemicals
J60045.MD |
250mg |
£44.52
250mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15440117
|
Thermo Scientific Chemicals
J60045.03 |
1g |
£156.00
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Inhibits α-melanocyte-stimulating hormone (α-MSH) release. Increases brain dopamine turnover and inhibits physiological and behavioral opioid effects in vivo. Role in vascular homeostasis.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
284.36 | |
Glass bottle | |
Lyophilized |
RUO – Research Use Only