missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Melon™ Gel Spin Plate Kit for IgG Screening

Rapidly purify human, mouse and rabbit IgG from serum and other sources with this simpler alternative to traditional Protein A and Protein G methods.

Brand:  Thermo Scientific™ 45208

Product Code. 11889360

  • £425.00 / Pack of 2

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

Thermo Scientific™ Melon Gel IgG Purification Resins and Kits comprise a simpler and faster alternative to traditional Protein A and Protein G methods for purification of human, mouse and rabbit IgG from serum and other sources.

Melon Gel IgG Purification Kits enable fast, easy and gentle purification of antibodies from serum and other sample types. Antibody purification with Melon Gel may be performed with gravity-flow columns, spin columns or chromatography cartridges. Melon Gel IgG Purification saves time compared to classical Protein A or Protein G methods without compromising the quality of the antibody. Compared to antibody affinity purification with Protein A or Protein G, Melon Gel IgG Purification is simpler, faster and provides greater yields and higher purity for a wider variety of antibodies species.

Highlights:

  • Simple, one-step protocol – IgG Purification with Melon Gel Resin does not require the binding, washing, and multiple elution steps required by Protein A and Protein G

  • Rapid purification – antibody purification from serum is four to six times faster than Protein A or G methods

  • High recovery and purity – antibody recovery from serum for many species is greater than 90% with greater than 80% purity

  • Minimal dilution – reduced need to concentrate antibody before storage or use

  • Robust purification – Melon Gel IgG Purification Kits work with a wide range of antibodies including many that do not purify well on Protein A or Protein G

  • Gentle purification – no harsh antibody elution conditions helps retain IgG activity

  • Reusable support – Melon Gel Resin can be used for multiple antibody purifications

  • Available in various formats – spin columns, purification kits and chromatography cartridges

TRUSTED_SUSTAINABILITY
Specifications

Specifications

Gel Spin Plate Kit
Proprietary Resin, Affinity
Proprietary Ligand
Melon Gel Spin Plate Kit for IgG Screening
2 Plates
Melon™
Kit, 96-well plate
IgG Antibodies
Kit Contents:
• Melon Gel 96-Well Spin Plates, 2 each, contains 100 μL of Melon Gel⁄well (Purification Capacity: 1 mg of human IgG⁄well)
• Melon Gel Purification Buffer, 2 × 100 mL
• Wash Plates (white), 2 each
• Collection Plate

Storage: Upon receipt store at 4°C. Product shipped at ambient temperature.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberAA397935 Certificate TypeCertificate of Analysis Date31/01/2025 Catalog Number45208
Lot NumberAA397935A Certificate TypeCertificate of Analysis Date31/01/2025 Catalog Number45208
Lot NumberZL393097 Certificate TypeCertificate of Analysis Date31/01/2025 Catalog Number45208
Lot NumberZI390088 Certificate TypeCertificate of Analysis Date16/10/2024 Catalog Number45208
Lot NumberZI390088A Certificate TypeCertificate of Analysis Date15/10/2024 Catalog Number45208
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Thermo Scientific™ Melon™ Gel Spin Plate Kit for IgG Screening > Filter Plates, 96-well, 100μL resin per well; 2 plates

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only. Not for use in diagnostic procedures.