Learn More
Thermo Scientific™ Melon™ Gel Monoclonal IgG Purification Kit
Shop All Thermo Scientific ProductsDescription
Thermo Scientific™ Melon Gel Monoclonal IgG Purification Kit is a high-yield, mild purification system for mouse monoclonal antibodies from serum, ascites or hybridoma cell culture supernatant.
Highlights:
Fast – purify antibodies in half the time of required for Protein A or G
Efficient – recovery and purity is often better than Protein A or G purification
Robust – effective for subclasses of IgG that bind poorly to Protein A or G
Gentle – antibodies retain activity because they are not exposed to denaturing conditions
Reusable – support can be regenerated and used for up to 6 purifications
Compatible – amine-free buffer does not require removal or neutralization
Flexible – accommodates a broad range of sample volumes and processing vessels
Includes:
Melon Gel Resin, purification buffer and resin regenerant
Specifications
Specifications
| Product Type | IgG Purification Kit |
| Product Line | Melon |
| Column Type | Proprietary Resin, Affinity |
| Format | Kit, Bottle |
| Stationary Phase | Proprietary Ligand |
| Purification Target | IgG Antibodies |
| Description | Melon Gel Monoclonal IgG Purification Kit |
| Content And Storage | Sufficient For: IgG purification from up to 1 L of cell culture supernatant or up to 200 mL of ascites fluid • Melon Gel Support, 200 mL • Melon Gel Purification Buffer (dry mix to make 100X), 1 L • Melon Gel Regenerant (dry mix to make 5X), 1 L Store at 4°C |
| Quantity | 200 mL |
For Research Use Only. Not for use in diagnostic procedures.
Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.