Learn More
BUCHI Melting and Boiling Point Instruments
Fast and accurate. BUCHI M-560/565 Melting Point Instruments have simple menus and programs that are easy to navigate on the intuitive LCD display, with either manual or automatic, real time video instrument.
£6265.00 - £10715.00
Product Code | Brand | Product Type | Includes | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Product Type | Includes | Price | Quantity & Availability | ||||
10499094
|
BUCHI
051999 |
Manual Melting Point Apparatus | Melting Point instrument |
£6265.00
Each |
Estimated Shipment: 06-08-2024
Log in to see stock. |
||||
12013303
|
BUCHI
51998 |
Automatic Melting Point Instrument | Melting Point instrument, Sample Loader M-569, Calibration Set M-565, melting point tubes (100), boiling point tubes (10), boiling capillary (10), sample holder, cleaning tool, CD demo melting point monitor, protective cover, USB cable (2m) |
£10715.00
Each |
Estimated Shipment: 06-08-2024
Log in to see stock. |
||||
Description
- Fail-safe calibration with automated determination and storage of calibration results
- Automatic determination of melting and boiling points
- Highly accurate temperature regulation and measurement
- Optimized reproducibility with homogeneous packing of the capillary using the Sample Loader M-569
- Control of almost any type of sample by measuring melting point, boiling point or slip melting point.
- Double check for difficult samples by comparing manual and automated measurement
- Observation and replay of the phase transition using color display and video recording
- PC software included for easy management of methods and results
- Compliant with Pharmacopeia methods (European, USP, Japanese)
- Comprehensive IQ/OQ procedures including certified calibration and verification compounds for GLP/GMP laboratories.