Learn More
Gibco™ MEM (10X), no glutamine
GREENER_CHOICE
MEM (10X), no glutamine
Brand:  Gibco™ 21430079
 View more versions of this product
Product Code. 11580566
  • £195.00 / 5000mL
Edge
Explore more special offers
For Research Use Only. All usage must comply with product instructions.
Description
Description
Minimum Essential Medium (MEM) is one of the most commonly used of all cell culture media. MEM can be used with a variety of suspension and adherent mammalian cells, including HeLa, BHK-21, 293, HEP-2, HT-1080, MCF-7, fibroblasts, and primary rat astrocytes. We offer a variety of Gibco™ MEM modifications for a range of cell culture applications. Find the right formulation using the media selector tool.
Gibco™ MEM, developed by Harry Eagle, was based on his earlier formulation of Basal Medium Eagle (BME). Many other modifications of MEM followed, including Glasgow’s MEM, MEM α, DMEM, and Temin’s Modification. MEM is available with Earle’s salts for use in a CO2 incubator, or with Hanks' salts for use without CO2. This product is made with Earle’s salts.
Product Intended Use
For in vitro diagnostic use. CAUTION: Not for human or animal therapeutic use. Uses other than the intended use may be a violation of local law.
cGMP Manufacturing and Quality System
Gibco™ MEM is manufactured at a cGMP compliant facility, located in Paisley, Scotland, UK. The facility is registered with the FDA as a medical device manufacturer and is certified to the ISO 13485 standard. For supply chain continuity, we offer a comparable Gibco™ MEM product made in our Grand Island facility. This facility is registered with the FDA as a medical device manufacturer and is certified to the ISO 13485 standards.
MEM contains no proteins, lipids, or growth factors. Therefore, MEM requires supplementation, commonly with 10% Fetal Bovine Serum (FBS). MEM uses a sodium bicarbonate buffer system (2.2 g/L) and therefore requires a 5–10% CO2 environment to maintain physiological pH.
Concentrated forms of Gibco™ cell culture medium require sodium bicarbonate supplementation, pH adjustment, and filtration at the time of preparation.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
ISO 13485 standards, FDA
MEM Medium, 500mL, 10X Concentration, Liquid Form, With Phenol Red, 1.2 to 1.8, 4.8 to 5.3 pH, 12 Months Shelf Life, 2400 to 3000mOsm/kg Osmolality, 2 to 8°C Storage Conditions, Used for Mammalian Cell Culture
Liquid
Phenol Red
Mammalian cell culture
10 x 500 mL
2 to 8°C
10 X
MEM Medium
No Glutamine, No HEPES, No Sodium Bicarbonate
1.2 to 1.8, 4.8 to 5.3
For in-vitro diagnostic use