missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ MEM Amino Acids Solution (50X)
GREENER_CHOICE

MEM Amino Acids Solution (50X)

Brand:  Gibco™ 11130036


 View more versions of this product

Product Code. 11570386

  • £18.42 / 100mL

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers

For Research Use Only. All usage must comply with product instructions.
 
This item is not returnable. View return policy

Description

Description

Gibco™ MEM Amino Acids Solution is used as a growth supplement for cell culture medium, to increase cell growth and viability. Gibco™ MEM Amino Acids Solution is formulated to contain 50X the essential amino acids (except L-glutamine) found in the standard Minimum Essential Medium (MEM). The complete formulation is available.

Product Use
For in vitro diagnostic use.

Dual-site cGMP Manufacturing and Quality System
Gibco™ MEM Amino Acids Solution is manufactured at a cGMP compliant facility, located in Paisley, Scotland, UK. The facility is registered with the FDA as a medical device manufacturer and is certified to the ISO 13485 standard. For supply chain continuity, we offer a comparable Gibco™ MEM Amino Acids Solution product made in our Grand Island facility (11130-051). This facility is registered with the FDA as a medical device manufacturer and is certified to the ISO 13485 standards.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

FDA, ISO 13485
MEM Amino Acids Solution, 50X Concentration, 100mL, Liquid, 0 to 2 pH, 2 to 8°C Storage Temperature, 12 Months Shelf Life, Room temperature and Ambient Shipping Condition, Used for Cell Culture, Mammalian Cell Culture
Liquid
Mammalian Cell Culture
100 mL
2 to 8°C
50 X
Amino Acid Supplement
0 to 2
RUO - research use only
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Product Identifier
  • MEM AMINO ACIDS W/O GLUT (50X)

Signal Word
  • Danger

Hazard Category
  • Skin corrosion/irritation Category 1 A
  • Serious eye damage/eye irritation Category 1

Hazard Statement
  • H314-Causes severe skin burns and eye damage.

Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

Supplemental information
  • MIXTURE LIST-Contains : Hydrogen chloride

SDS
Documents

Documents

Certificates
Special Offers

Special Offers