missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Gibco™ MEM, NEAA, no glutamine

MEM, NEAA, no glutamine
£32.05 - £292.00
Specifications
Form | Liquid |
---|---|
Product Type | Minimum Essential Medium NEAA |
With Additives | Non Essential Amino Acids (NEAA), Phenol Red |
Without Additives | No Glutamine, No HEPES |
For Use With (Application) | For cell culture, mammalian cell culture, In-vitro cultivation |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11583397
![]() |
Gibco™
10370047 |
500 mL |
£32.05
500mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11593397
![]() |
Gibco™
10370070 |
10 x 500 mL |
£292.00
Pack of 10 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Minimum Essential Medium (MEM) is one of the most commonly used of all cell culture media. MEM can be used with a variety of suspension and adherent mammalian cells, including HeLa, BHK-21, 293, HEP-2, HT-1080, MCF-7, fibroblasts, and primary rat astrocytes. We offer a variety of Gibco™ MEM modifications for a range of cell culture applications. Find the right formulation using the media selector tool.Gibco™ MEM, developed by Harry Eagle, was based on his earlier formulation of Basal Medium Eagle (BME). Many other modifications of MEM followed, including Glasgow's MEM, MEM α, DMEM, and Temin's Modification. MEM is available with Earle's salts for use in a CO2 incubator, or with Hanks' salts for use without CO2. This product is made with Earle's salts.
Product Intended Use
For in vitro diagnostic use. CAUTION: Not for human or animal therapeutic use. Uses other than the intended use may be a violation of local law.
Dual-Site cGMP Manufacturing
Gibco™ MEM is manufactured at a cGMP compliant facility, located in Grand Island, New York. The facility is registered with the FDA as a medical device manufacturer and is certified to ISO 13485 standards. For supply chain continuity, we offer an identical Gibco™ MEM product made in our Scotland facility. This facility is registered with the FDA as a medical device manufacturer and is certified to the ISO 13485 standard.
MEM contains no proteins, lipids, or growth factors. Therefore, MEM requires supplementation, commonly with 10%Fetal Bovine Serum (FBS). MEM uses a sodium bicarbonate buffer system (2.2 g/L) and therefore requires a 5–10% CO2 environment to maintain physiological pH.
Specifications
Liquid | |
Non Essential Amino Acids (NEAA), Phenol Red | |
For cell culture, mammalian cell culture, In-vitro cultivation | |
2 to 8°C |
Minimum Essential Medium NEAA | |
No Glutamine, No HEPES | |
ISO 13485 standards, FDA |
Safety and Handling
missing translation for 'shelfLife' : 12 Months
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Gibco™ MEM, NEAA, no glutamine