Learn More
Gibco™ MEM Non-Essential Amino Acids Solution (100X)
Gibco™ MEM Non-Essential Amino Acids are used as a supplement for cell culture medium, to increase cell growth and viability.
Brand: Gibco™ 11140050
15.64 GBP valid until 2025-02-21
Use promo code "24115" to get your promotional price.
Description
Gibco™ MEM Non-Essential Amino Acids are used as a supplement for cell culture medium, to increase cell growth and viability. Gibco™ MEM Non-Essential Amino Acids contains the same non-essential amino acids found in the standard Minimum Essential Medium (MEM) at a strength of 100X. The complete formulation is available.
Specifications
Store at 2–8°C. Protect from light. | |
100X | |
Amino Acid Supplement | |
cGMP-compliant under the ISO 13485 standard | |
100 mL |
Mammalian Cells | |
Liquid | |
Sterile-filtered | |
0.9 | |
Room Temperature |
Safety and Handling
- MEM NON-ESSENTIAL AMINO ACIDS SOLUTION, (100X)
Signal Word
- Warning
Hazard Category
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 2
Hazard Statement
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
Precautionary Statement
- P264-Wash hands thoroughly after handling.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/soap.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P332+P313-If skin irritation occurs: Get medical advice/attention.
- P337+P313-If eye irritation persists: Get medical advice/attention.
- P362+P364-Take off contaminated clothing and wash it before reuse.
Supplemental information
- MIXTURE LIST-Contains : amino acids
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use or Further Manufacturing. Not for diagnostic use or direct administration into humans or animals.