missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Gibco™ MEM α, nucleosides
MEM α, nucleosides
Brand: Gibco™ 22571038
421.65 GBP valid until 2024-12-31
Use promo code "22915" to get your promotional price.
Code : 50
Additional Details : Weight : 0.50000kg
Description
Minimum Essential Medium (MEM) α is widely used for mammalian cell culture as well as selection for transfected DHFR negative cells. MEM α can be used with a variety of suspension and adherent mammalian cells, including keratinocytes, primary rat astrocytes, and human melanoma cells. We offer a variety of Gibco™ MEM α modifications for a range of cell culture applications. Find the right formulation using the media selector tool.Gibco™ MEM α is a modification of Minimum Essential Medium (MEM) that contains non-essential amino acids, sodium pyruvate, lipoic acid, B12, biotin, and ascorbic acid. MEM α is available without nucleosides for use as a selection medium for DG44 and other DHFR negative cells. This product is made with Earle’s salts.
Product Intended Use
For in vitro diagnostic use. CAUTION: Not for human or animal therapeutic use. Uses other than the intended use may be a violation of local law.
cGMP Manufacturing and Quality System
Gibco™ MEM α is manufactured at a cGMP compliant facility, located in Paisley, Scotland, UK. The facility is registered with the FDA as a medical device manufacturer and is certified to the ISO 13485 standard. For supply chain continuity, we offer an identical Gibco™ MEM α product made in our Grand Island facility. This facility is registered with the FDA as a medical device manufacturer and is certified to the ISO 13485 standards.
MEM α contains no proteins, lipids, or growth factors. Therefore, MEM α requires supplementation, commonly with 10%Fetal Bovine Serum (FBS). MEM α uses a sodium bicarbonate buffer system (2.2 g/L) and therefore requires a 5-10% CO2 environment to maintain physiological pH.
Specifications
ISO 13485 standards, FDA | |
MEM α± Medium, 500mL, 1 X Concentration, Liquid Form, With Ribonucleosides, Deoxyribonucleosides, L-glutamine, Phenol Red, Sodium Pyruvate, 7 to 7.4 pH, 12 Months Shelf Life, 280 to 310mOsm/kg Osmolality, Used for Mammalian Cell Culture | |
Liquid | |
Glutamine, Phenol Red, Sodium Pyruvate, Ribonucleosides, Deoxyribonucleosides | |
7 to 7.4 | |
For in-vitro diagnostic use |
2 to 8°C | |
1 X | |
MEM α± Medium | |
Mammalian cell culture | |
10 x 500 mL |