missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Gibco™ MEM α, nucleosides, GlutaMAX™ Supplement
MEM α, nucleosides, GlutaMAX™ Supplement
£75.75 - £650.00
Specifications
Certifications/Compliance | ISO 13485 standards, FDA |
---|---|
Content And Storage | 2 to 8°C |
Concentration | 1 X |
Form | Liquid |
Product Type | Minimum Essential Medium α± |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11544466
|
Gibco™
32571028 |
500 mL |
£75.75
500mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11554466
|
Gibco™
32571093 |
10 x 500 mL |
£650.00
Pack of 10 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Minimum Essential Medium (MEM) α is widely used for mammalian cell culture as well as selection for transfected DHFR negative cells. MEM α can be used with a variety of suspension and adherent mammalian cells, including keratinocytes, primary rat astrocytes, and human melanoma cells. We offer a variety of Gibco™ MEM α modifications for a range of cell culture applications. Find the right formulation using the media selector tool.Gibco™ MEM α is a modification of Minimum Essential Medium (MEM) that contains non-essential amino acids, sodium pyruvate, lipoic acid, B12, biotin, and ascorbic acid. MEM α is available without nucleosides for use as a selection medium for DG44 and other DHFR negative cells. Gibco™ MEM α with GlutaMAX™ supplement minimizes toxic ammonia build-up and improves cell viability and growth in an easy-to-use format. This product is made with Earle’s salts.
Product Intended Use
For in vitro diagnostic use. CAUTION: Not for human or animal therapeutic use. Uses other than the intended use may be a violation of local law.
cGMP Manufacturing and Quality System
Gibco™ MEM α is manufactured at a cGMP compliant facility, located in Paisley, Scotland, UK. The facility is registered with the FDA as a medical device manufacturer and is certified to the ISO 13485 standard. For supply chain continuity, we offer an identical Gibco™ MEM α product made in our Grand Island facility. This facility is registered with the FDA as a medical device manufacturer and is certified to the ISO 13485 standards.
MEM α contains no proteins, lipids, or growth factors. Therefore, MEM α requires supplementation, commonly with 10% Fetal Bovine Serum (FBS). MEM α uses a sodium bicarbonate buffer system (2.2 g/L) and therefore requires a 5-10% CO2 environment to maintain physiological pH.
Specifications
ISO 13485 standards, FDA | |
1 X | |
Minimum Essential Medium α± | |
7.0 to 7.4 |
2 to 8°C | |
Liquid | |
Mammalian cell culture |
Safety and Handling
missing translation for 'shelfLife' : 12 Months
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title