missing translation for 'onlineSavingsMsg'
Learn More

Memantine hydrochloride, Tocris Bioscience™

Product Code. 30001128
Click to view available options
Quantity:
50 mg
This item is not returnable. View return policy
This item is not returnable. View return policy

NMDA antagonist; acts at ion channel site

Memantine hydrochloride is an an antagonist at the NMDA receptor, binding to the ion channel site. Used in the treatment of Parkinsonism.

Specifications

CAS 41100-52-1
Molecular Formula C12H22ClN
MDL Number MFCD00214336
Quantity 50 mg
Synonym memantine hydrochloride, memantine hcl, 3,5-dimethyladamantan-1-amine hydrochloride, 3,5-dimethyl-1-adamantanamine hydrochloride, namenda, akatinol, axura, namenda xr, memantine.hcl
InChI Key LDDHMLJTFXJGPI-UHFFFAOYNA-N
SMILES [H+].[Cl-].CC12CC3CC(C)(C1)CC(N)(C3)C2
IUPAC Name hydrogen 3,5-dimethyladamantan-1-amine chloride
Molecular Weight (g/mol) 215.77
PubChem CID 181458
ChEBI CHEBI:64323
Formula Weight 215.77
Percent Purity >98%
Chemical Name or Material Memantine hydrochloride
Show More Show Less
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'