missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Membrane Lactose Glucoronide Agar (MLGA)

Product Code. 10422083
Click to view available options
Quantity:
500 g
Unit Size:
500g
This item is not returnable. View return policy

Product Code. 10422083

Brand: Thermo Scientific™ CM1031B

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Detect, enumerate, and differentiate Escherichia coli and other coliforms by single membrane filtration with this selective and differential medium.

Detect, enumerate and differentiate Escherichia coli and other coliforms by single membrane filtration with Thermo Scientific™ Oxoid™ Membrane Lactose Glucuronide Agar (MLGA) (Dehydrated). The presence of lauryl sulphate and lactose with phenol red allow the selective isolation and differentiation of Escherichia coli and coliforms respectively.

Tests for Escherichia coli and coliforms provide a sensitive means for detection of fecal contamination of drinking water. They are regarded as the most important routine microbial examinations for assessing the effectiveness of water treatment and disinfection and for monitoring water quality in distribution1. Membrane Lactose Glucuronide Agar (MLGA) supports water testing utilising single membrane filtration.
  • Isolate coliforms effectively with MLGA using a single membrane filtration method.
  • Lauryl sulphate selects for coliforms by inhibiting Gram-positive organisms.
  • Reduced confirmatory tests for E. coli due to the specificity of the reactions in the medium.
  • Easy interpretation of the test results due to addition of chromogenic BCIG (5-bromo-4-chloro-3-indolyl-ß-D-glucuronide), E. coli appear as green colonies while other coliform bacteria form yellow colonies.
Identification of Escherichia coli and coliforms is facilitated through two biochemical reactions within the medium: Lactose fermentation is detected by the dye phenol red which gives yellow colonies when acid is produced. The chromogenic substrate BCIG is cleaved by the enzyme glucuronidase and produces a blue chromophore which builds up in the bacterial cells. Coliforms are lactose-positive so colonies will be yellow; Escherichia coli is both lactose-positive and possesses glucuronidase so will appear as green colonies.1

Not all products are available for sale in all territories. Please inquire.
Remel™ and Oxoid™ products are now part of the Thermo Scientific™ brand.
TRUSTED_SUSTAINABILITY

Specifications

Content And Storage 10°C to 30°C
Description Membrane Lactose Glucuronide Agar (MLGA)(Dehydrated)
Form Powder
Product Type Agar
Quantity 500 g
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.