Learn More
Fisherbrand™ Ion Selective Membrane Replacement Kits
Sample preparation steps such as filtering and distillation are rarely necessary with this kit for the combined ion-sensitive electrode with a solid membrane.
Brand:  Fisherbrand™ CALMK22
 View more versions of this product
Product Code. 11719728
  • £297.00 / Each
Estimated Shipment: 02-05-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Membrane replacement kit for ion-sensitive electrodes
  • For  83919 device
  • Allows for analysis time generally less than one minute
Specifications
Specifications
Membrane replacement kit for ion-sensitive electrodes.
83919 Device
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Fisherbrand™ Ion Selective Membrane Replacement Kits > For Use With: 83919